siRNA Details
| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029961.2 | NP_084237.1 | 25 | 3133 | ||
| GenBank mRNA | AY766239.1 | - | 25 | 3097 | ||
| JQ655148.1 | - | 12 | 2612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb5| Gene Symbol: Abcb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029961.2 | NP_084237.1 | 25 | 3133 | ||
| GenBank mRNA | AY766239.1 | - | 25 | 3097 | ||
| JQ655148.1 | - | 12 | 2612 | |||