siRNA Details
| Gene Symbol: Ermn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029972.3 | NP_084248.2 | 3 | 805 | ||
| GenBank mRNA | AK078089.1 | - | 3 | 893 | ||
| AK136820.1 | - | 3 | 895 | |||
| AK137305.1 | - | 3 | 897 | |||
| AK220372.1 | - | 3 | 927 | |||
| BC089515.1 | - | 3 | 806 | 6589784 | ||
| BC090670.1 | - | 3 | 837 | 30941185 | ||
| DQ334270.1 | - | 3 | 783 | |||

Ermn
Genomic Map