siRNA Details
| Gene Symbol: Epm2aip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175266.4 | NP_780475.1 | 1 | 899 | ||
| GenBank mRNA | AK030084.1 | - | 1 | 816 | ||
| AK032284.1 | - | 1 | 870 | |||
| AK043518.1 | - | 1 | 858 | |||
| BC018474.1 | - | 1 | 903 | 3156440 | ||

Epm2aip1
Genomic Map