siRNA Details
| Gene Symbol: TONSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013432.4 | NP_038460.4 | 6 | 608 | ||
| XM_011517050.2 | XP_011515352.1 | 6 | 667 | |||
| GenBank mRNA | AB209126.1 | - | 1 | 459 | ||
| BC008782.2 | - | 5 | 580 | 3628374 | ||

TONSL
Genomic Map