siRNA Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081222.1 | NP_001074691.1 | 11 | 3431 | ||
| GenBank mRNA | AK040576.1 | - | 11 | 1259 | ||
| AK049589.1 | - | 10 | 1148 | |||
| AK145333.1 | - | 11 | 3432 | |||
| AK145870.1 | - | 10 | 1079 | |||
| AK173297.1 | - | 9 | 2809 | |||
| BC008220.1 | - | 10 | 1303 | |||
| BC076613.1 | - | 11 | 3161 | |||
| BC151069.1 | - | 11 | 3387 | 9087982 | ||
| BC151077.1 | - | 11 | 3387 | 9087990 | ||
| BC172153.1 | - | 12 | 3509 | |||

Esco1
Genomic Map