siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 9 | 1460 | ||
| GenBank mRNA | AK040525.1 | - | 9 | 1462 | ||
| AK128931.1 | - | 9 | 1411 | |||
| AK162934.1 | - | 9 | 1168 | |||
| AK173281.1 | - | 9 | 3164 | |||
| BC066816.1 | - | 9 | 1380 | 30084911 | ||

Krba1
Genomic Map