siRNA Details
| Gene Symbol: Thyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144543.2 | NP_653126.1 | 3 | 207 | ||
| GenBank mRNA | AB080370.1 | - | 2 | 131 | ||
| AK021335.1 | - | 3 | 175 | |||
| AK077639.1 | - | 3 | 177 | |||
| BC016594.1 | - | 3 | 161 | 2631627 | ||
| CB236103.1 | - | 2 | 400 | |||

Thyn1
Genomic Map