siRNA Details
| Gene Symbol: E130102H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI853970.1 | - | 1, 2 | 170 | ||
| AK021368.1 | - | 1, 2 | 132 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130102H24Rik| Gene Symbol: E130102H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI853970.1 | - | 1, 2 | 170 | ||
| AK021368.1 | - | 1, 2 | 132 | |||