siRNA Details
| Gene Symbol: 6030498E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183126.2 | NP_898949.2 | 3 | 530 | ||
| GenBank mRNA | AK020088.1 | - | 3 | 532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6030498E09Rik| Gene Symbol: 6030498E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183126.2 | NP_898949.2 | 3 | 530 | ||
| GenBank mRNA | AK020088.1 | - | 3 | 532 | ||