siRNA Details
| Gene Symbol: Lbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029999.4 | NP_084275.3 | 3 | 523 | ||
| GenBank mRNA | AF317517.1 | - | 3 | 506 | ||
| AK020110.1 | - | 3 | 523 | |||
| BC026827.1 | - | 3 | 505 | 4218560 | ||
| BC052470.1 | - | 3 | 503 | 6813866 | ||

Lbh
Genomic Map