siRNA Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004363.1 | NP_001004363.1 | 7 | 1291 | ||
| GenBank mRNA | AB182364.1 | - | 6 | 928 | ||
| BC040467.1 | - | 3 | 587 | |||
| BC082328.1 | - | 7 | 1291 | 30355405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nuak1| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004363.1 | NP_001004363.1 | 7 | 1291 | ||
| GenBank mRNA | AB182364.1 | - | 6 | 928 | ||
| BC040467.1 | - | 3 | 587 | |||
| BC082328.1 | - | 7 | 1291 | 30355405 | ||