siRNA Details
| Gene Symbol: Ccdc150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030025.2 | NP_084301.2 | 3 | 491 | ||
| GenBank mRNA | AK019675.1 | - | 3 | 489 | ||
| AK029994.1 | - | 3 | 491 | |||
| BC145597.1 | - | 3 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc150| Gene Symbol: Ccdc150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030025.2 | NP_084301.2 | 3 | 491 | ||
| GenBank mRNA | AK019675.1 | - | 3 | 489 | ||
| AK029994.1 | - | 3 | 491 | |||
| BC145597.1 | - | 3 | 452 | |||