siRNA Details
| Gene Symbol: Ccdc150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030025.2 | NP_084301.2 | 21 | 2476 | ||
| GenBank mRNA | AK029994.1 | - | 21 | 2476 | ||
| BC145597.1 | - | 23 | 2566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc150| Gene Symbol: Ccdc150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030025.2 | NP_084301.2 | 21 | 2476 | ||
| GenBank mRNA | AK029994.1 | - | 21 | 2476 | ||
| BC145597.1 | - | 23 | 2566 | |||