siRNA Details
| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030028.1 | NP_084304.1 | 5 | 389 | ||
| GenBank mRNA | AK019807.1 | - | 5 | 382 | ||
| BU936271.1 | - | 392 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem190| Gene Symbol: Tmem190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030028.1 | NP_084304.1 | 5 | 389 | ||
| GenBank mRNA | AK019807.1 | - | 5 | 382 | ||
| BU936271.1 | - | 392 | ||||