siRNA Details
| Gene Symbol: Cpt1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252470.1 | NP_001239399.1 | 3 | 515 | ||
| NM_153679.2 | NP_710146.1 | 3 | 517 | |||
| GenBank mRNA | AF320000.1 | - | 3 | 440 | ||
| AK035790.1 | - | 3 | 481 | |||
| AK087747.1 | - | 3 | 389 | |||
| BC066155.1 | - | 3 | 439 | 30089333 | ||
| BC083317.1 | - | 3 | 398 | |||
| CN842042.1 | - | 520 | ||||

Cpt1c
Genomic Map