siRNA Details
| Gene Symbol: Spag11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153115.1 | NP_694755.1 | 2 | 372 | ||
| GenBank mRNA | AB089183.1 | - | 2 | 372 | ||
| AF384173.1 | - | 2 | 335 | |||
| AK020333.1 | - | 2 | 376 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag11a| Gene Symbol: Spag11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153115.1 | NP_694755.1 | 2 | 372 | ||
| GenBank mRNA | AB089183.1 | - | 2 | 372 | ||
| AF384173.1 | - | 2 | 335 | |||
| AK020333.1 | - | 2 | 376 | |||