siRNA Details
| Gene Symbol: Tmem210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030055.1 | NP_084331.1 | 1 | 91 | ||
| GenBank mRNA | AK019803.1 | - | 1 | 92 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem210| Gene Symbol: Tmem210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030055.1 | NP_084331.1 | 1 | 91 | ||
| GenBank mRNA | AK019803.1 | - | 1 | 92 | ||