siRNA Details
| Gene Symbol: Spink12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030061.3 | NP_084337.1 | 2, 3 | 283 | ||
| GenBank mRNA | AK020352.1 | - | 2, 3 | 184 | ||
| AK136616.1 | - | 2, 3 | 284 | |||
| BC145914.1 | - | 2, 3 | 169 | 40131217 | ||
| BC145916.1 | - | 2, 3 | 169 | 40131219 | ||
| DQ437332.1 | - | 2, 3 | 342 | |||

Spink12
Genomic Map