siRNA Details
| Gene Symbol: Adgrf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289499.1 | NP_001276428.1 | 6 | 987 | ||
| NM_001289500.1 | NP_001276429.1 | 6 | 974 | |||
| NM_001289501.1 | NP_001276430.1 | 6 | 1018 | |||
| NM_030067.2 | NP_084343.1 | 6 | 1061 | |||
| GenBank mRNA | AK019508.1 | - | 6 | 1064 | ||
| BC089564.1 | - | 6 | 987 | 30502915 | ||
| BC140980.1 | - | 6 | 927 | 9055592 | ||

Adgrf4
Genomic Map