siRNA Details
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111016.1 | NP_001104486.1 | 8 | 2465 | ||
| NM_175272.3 | NP_780481.2 | 7 | 2727 | |||
| GenBank mRNA | AK040152.1 | - | 6 | 2012 | ||
| AK129480.1 | - | 7 | 2022 | |||
| BC038177.1 | - | 2 | 1156 | |||
| BC150679.1 | - | 6 | 1715 | |||

Nav2
Genomic Map