siRNA Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF376726.1 | - | 11 | 1120 | ||
| AK019012.1 | - | 2 | 94 | |||
| AK047364.1 | - | 11 | 1096 | |||
| AK082797.1 | - | 11 | 1146 | |||
| AK159313.1 | - | 11 | 1165 | |||
| AK170948.1 | - | 10 | 1248 | |||
| AK171030.1 | - | 10 | 1248 | |||
| AX775895.1 | - | 11 | 1177 | |||
| AX775897.1 | - | 11 | 1120 | |||
| BC016104.1 | - | 11 | 1146 | 4920840 | ||

Gpr108
Genomic Map