siRNA Details
| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081291.1 | NP_001074760.1 | 4 | 417 | ||
| GenBank mRNA | BC151001.1 | - | 4 | 390 | 9087914 | |
| BC151009.1 | - | 4 | 390 | 9087922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc88b| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081291.1 | NP_001074760.1 | 4 | 417 | ||
| GenBank mRNA | BC151001.1 | - | 4 | 390 | 9087914 | |
| BC151009.1 | - | 4 | 390 | 9087922 | ||