siRNA Details
| Gene Symbol: Ccdc88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081291.1 | NP_001074760.1 | 14 | 2681 | ||
| GenBank mRNA | BC076600.1 | - | 5 | 1565 | ||
| BC151001.1 | - | 14 | 2654 | 9087914 | ||
| BC151009.1 | - | 14 | 2654 | 9087922 | ||
| BC157946.1 | - | 14 | 2654 | 9088122 | ||

Ccdc88b
Genomic Map