siRNA Details
| Gene Symbol: Ttyh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052340.1 | - | 10 | 1250 | ||
| AK173233.1 | - | 7 | 639 | |||
| BC059083.1 | - | 10 | 1250 | 6827289 | ||
| BC062917.1 | - | 10 | 1256 | 6831835 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttyh3| Gene Symbol: Ttyh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052340.1 | - | 10 | 1250 | ||
| AK173233.1 | - | 7 | 639 | |||
| BC059083.1 | - | 10 | 1250 | 6827289 | ||
| BC062917.1 | - | 10 | 1256 | 6831835 | ||