siRNA Details
| Gene Symbol: Fam131a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133778.2 | NP_598539.1 | 6 | 941 | ||
| GenBank mRNA | AK049983.1 | - | 6 | 942 | ||
| BC003957.1 | - | 1 | 275 | |||
| BC092532.1 | - | 6 | 972 | 6594874 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam131a| Gene Symbol: Fam131a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133778.2 | NP_598539.1 | 6 | 941 | ||
| GenBank mRNA | AK049983.1 | - | 6 | 942 | ||
| BC003957.1 | - | 1 | 275 | |||
| BC092532.1 | - | 6 | 972 | 6594874 | ||