siRNA Details
| Gene Symbol: 3110062M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135611.1 | NP_001129083.1 | 2 | 329 | ||
| NM_199145.2 | NP_954596.1 | 2 | 654 | |||
| GenBank mRNA | AK042380.1 | - | 2 | 610 | ||
| AK083365.1 | - | 2 | 296 | |||
| AK170119.1 | - | 2 | 325 | |||
| BB611813.1 | - | 2 | 330 | |||
| BC029235.1 | - | 2 | 273 | 4912211 | ||

3110062M04Rik
Genomic Map