siRNA Details
| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277980.1 | NP_001264909.1 | 2 | 268 | ||
| NM_030104.1 | NP_084380.1 | 2 | 268 | |||
| GenBank mRNA | AK018916.1 | - | 2 | 202 | ||
| AK076910.1 | - | 2 | 179 | |||
| BC100486.1 | - | 2 | 182 | |||
| CN837600.1 | - | 269 | ||||

Tmem247
Genomic Map