siRNA Details
| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277980.1 | NP_001264909.1 | 1 | 66 | ||
| NM_030104.1 | NP_084380.1 | 1 | 66 | |||
| GenBank mRNA | CN837600.1 | - | 67 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem247| Gene Symbol: Tmem247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277980.1 | NP_001264909.1 | 1 | 66 | ||
| NM_030104.1 | NP_084380.1 | 1 | 66 | |||
| GenBank mRNA | CN837600.1 | - | 67 | |||