siRNA Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030113.2 | NP_084389.2 | 17, 18 | 1677 | ||
| GenBank mRNA | AF297029.1 | - | 17, 18 | 1581 | ||
| AF297030.1 | - | 17, 18 | 1640 | |||
| AY179965.1 | - | 17, 18 | 1553 | |||
| AY179966.1 | - | 16, 17 | 1487 | |||
| AY179967.1 | - | 17, 18 | 1553 | |||
| BU151977.1 | - | 7, 8 | 503 | |||

Arhgap10
Genomic Map