siRNA Details
| Gene Symbol: Arhgap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030113.2 | NP_084389.2 | 22 | 2363 | ||
| GenBank mRNA | AF297029.1 | - | 22 | 2267 | ||
| AF297030.1 | - | 22 | 2326 | |||
| AY179965.1 | - | 22 | 2239 | |||
| AY179967.1 | - | 21 | 2086 | |||
| BI736055.1 | - | 2 | 147 | |||

Arhgap10
Genomic Map