siRNA Details
| Gene Symbol: Adgra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054044.2 | NP_473385.2 | 19 | 3062 | ||
| GenBank mRNA | AF378759.1 | - | 19 | 3062 | ||
| BC138451.1 | - | 19 | 3052 | 8861471 | ||
| BC138452.1 | - | 19 | 3052 | 8861472 | ||
More Information
Set Membership: |
|

Adgra3
Genomic Map