siRNA Details
| Gene Symbol: Utp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030132.5 | NP_084408.1 | 1 | 131 | ||
| GenBank mRNA | AK021306.1 | - | 1 | 81 | ||
| AK042518.1 | - | 1 | 88 | |||
| AK079831.1 | - | 1 | 99 | |||
| AK134445.1 | - | 1 | 63 | |||
| BC027540.1 | - | 1 | 67 | |||
| BC046791.1 | - | 1 | 76 | 5717427 | ||
| BC056961.1 | - | 1 | 76 | 5717427 | ||
| CK334116.1 | - | 1 | 143 | |||

Utp23
Genomic Map