siRNA Details
Gene Symbol: 8030411F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030135.2 | NP_084411.1 | 1 | 185 | ||
GenBank mRNA | AF440735.1 | - | 1 | 185 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8030411F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030135.2 | NP_084411.1 | 1 | 185 | ||
GenBank mRNA | AF440735.1 | - | 1 | 185 |