siRNA Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084820.1 | - | 2 | 1607 | ||
| AK160104.1 | - | 2 | 1576 | |||
| AK170084.1 | - | 2 | 1589 | |||
| BC147206.1 | - | 1 | 191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tulp4| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084820.1 | - | 2 | 1607 | ||
| AK160104.1 | - | 2 | 1576 | |||
| AK170084.1 | - | 2 | 1589 | |||
| BC147206.1 | - | 1 | 191 | |||