siRNA Details
Gene Symbol: Eif3j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256055.1 | NP_001242984.1 | 1 | 444 | ||
GenBank mRNA | BC080788.1 | - | 1 | 398 | 4166214 | |
BC108355.1 | - | 1 | 224 |
Gene Symbol: Eif3j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144545.4 | NP_653128.2 | 5 | 441 | ||
GenBank mRNA | AK082048.1 | - | 4 | 232 | ||
AK147154.1 | - | 5 | 373 | |||
AK148004.1 | - | 5 | 444 | |||
AK154934.1 | - | 5 | 442 | |||
AK161038.1 | - | 5 | 399 | |||
BC006055.1 | - | 1 | 18 |
