siRNA Details
| Gene Symbol: Plekhj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023900.2 | NP_076389.2 | 4 | 706 | ||
| GenBank mRNA | AK020621.1 | - | 4 | 375 | ||
| AK051734.1 | - | 4 | 363 | |||
| AK088567.1 | - | 4 | 669 | |||
| AK150144.1 | - | 4 | 321 | |||
| AK178993.1 | - | 2 | 111 | |||
| AK219000.1 | - | 2 | 43 | |||
| BC005565.1 | - | 4 | 304 | 3966305 | ||

Plekhj1
Genomic Map