siRNA Details
| Gene Symbol: Plekhj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023900.2 | NP_076389.2 | 6 | 882 | ||
| GenBank mRNA | AK020621.1 | - | 6 | 551 | ||
| AK088567.1 | - | 6 | 845 | |||
| AK150144.1 | - | 6 | 497 | |||
| AK178993.1 | - | 4 | 287 | |||
| AK219000.1 | - | 4 | 219 | |||
| BC005565.1 | - | 6 | 480 | 3966305 | ||

Plekhj1
Genomic Map