siRNA Details
| Gene Symbol: 9530057J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020613.1 | - | 833 | |||
| Gene Symbol: Gm3643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029875.1 | - | 3055 | |||

9530057J20Rik
Genomic Map