siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 14 | 1291 | ||
| GenBank mRNA | AK050805.1 | - | 4 | 608 | ||
| AK191882.1 | - | 287 | ||||
| AY102701.1 | - | 14 | 1213 | |||
| BC056435.1 | - | 14 | 1200 | 5707801 | ||

Naa35
Genomic Map