siRNA Details
| Gene Symbol: Naa35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030153.2 | NP_084429.2 | 18 | 1786 | ||
| GenBank mRNA | AK039339.1 | - | 1 | 317 | ||
| AK052852.1 | - | 1 | 252 | |||
| AY102701.1 | - | 18 | 1708 | |||
| BC010475.1 | - | 1 | 84 | |||
| BC056435.1 | - | 18 | 1695 | 5707801 | ||

Naa35
Genomic Map