siRNA Details
| Gene Symbol: Pus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289780.1 | NP_001276709.1 | 2 | 431 | ||
| NM_001289781.1 | NP_001276710.1 | 2 | 782 | |||
| NM_178403.5 | NP_848490.2 | 2 | 431 | |||
| GenBank mRNA | AK142271.1 | - | 2 | 400 | ||
| AK145380.1 | - | 2 | 782 | |||
| AK160849.1 | - | 2 | 383 | |||
| AK173290.1 | - | 2 | 371 | |||
| BC008544.1 | - | 2 | 385 | 3497634 | ||
| BC138533.1 | - | 2 | 273 | 8861553 | ||
| BC145316.1 | - | 2 | 273 | 9053871 | ||

Pus7
Genomic Map