siRNA Details
Gene Symbol: Galnt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030166.3 | NP_084442.1 | 5 | 1512 | ||
GenBank mRNA | AB175682.1 | - | 5 | 1122 | ||
AK019470.1 | - | 5 | 1627 | |||
AK030347.1 | - | 5 | 765 | |||
AK050075.1 | - | 5 | 1012 | |||
AK090330.1 | - | 3 | 835 | |||
AK163883.1 | - | 5 | 1388 | |||
BC133711.1 | - | 5 | 1512 | 40143107 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008771060.
Panther Classification:
Gene Ontology Categories:
