siRNA Details
| Gene Symbol: Fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166365.2 | NP_001159837.2 | 4, 5 | 1110 | ||
| NM_001166583.1 | NP_001160055.1 | 4, 5 | 1110 | |||
| NM_030167.4 | NP_084443.1 | 4, 5 | 1110 | |||
| GenBank mRNA | AB541013.1 | - | 4, 5 | 278 | ||
| AK019480.1 | - | 4, 5 | 337 | |||
| AK036417.1 | - | 4, 5 | 1107 | |||
| AK044896.1 | - | 4, 5 | 1111 | |||

Fam122b
Genomic Map