siRNA Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 20 | 2302 | ||
| GenBank mRNA | AK013379.1 | - | 7 | 908 | ||
| AK047562.1 | - | 20 | 2303 | |||
| BB197311.2 | - | 2 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mctp1| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030174.2 | NP_084450.2 | 20 | 2302 | ||
| GenBank mRNA | AK013379.1 | - | 7 | 908 | ||
| AK047562.1 | - | 20 | 2303 | |||
| BB197311.2 | - | 2 | 237 | |||