siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183137.2 | NP_898960.2 | 2 | 163 | ||
| GenBank mRNA | AK019105.1 | - | 2 | 134 | ||
| AK154660.1 | - | 2 | 128 | |||
| AK202725.1 | - | 2 | 78 | |||
| AK214511.1 | - | 2 | 78 | |||
| BB868415.1 | - | 2 | 164 | |||
| BC094318.1 | - | 2 | 113 | |||
| BC137580.1 | - | 2 | 132 | 8860599 | ||
| BC144724.1 | - | 2 | 132 | 9053272 | ||
| BY084429.1 | - | 2 | 133 | |||

Enthd2
Genomic Map