siRNA Details
| Gene Symbol: Enthd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183137.2 | NP_898960.2 | 12, 13 | 1292 | ||
| GenBank mRNA | AK019105.1 | - | 12, 13 | 1264 | ||
| AK154660.1 | - | 12, 13 | 1257 | |||
| BC049110.1 | - | 12, 13 | 1228 | |||
| BC064313.1 | - | 12, 13 | 1228 | |||
| BC094318.1 | - | 11, 12 | 1781 | |||
| BC137580.1 | - | 12, 13 | 1261 | 8860599 | ||
| BC144724.1 | - | 12, 13 | 1261 | 9053272 | ||

Enthd2
Genomic Map