siRNA Details
| Gene Symbol: Zc3hav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028421.1 | NP_082697.1 | 8, 9 | 2637 | ||
| NM_028864.2 | NP_083140.1 | 8, 9 | 2637 | |||
| GenBank mRNA | AK004770.1 | - | 8, 9 | 2638 | ||
| AK020263.2 | - | 8, 9 | 2316 | |||
| AK080334.1 | - | 8, 9 | 2622 | |||
| AK143568.1 | - | 8, 9 | 2307 | |||
| AK169402.1 | - | 8, 9 | 2282 | |||
| AK172379.1 | - | 8, 9 | 2638 | |||
| BC029090.1 | - | 8, 9 | 2199 | 2651558 | ||

Zc3hav1
Genomic Map