siRNA Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282126.1 | NP_001269055.1 | 3 | 1542 | ||
| NM_001282127.1 | NP_001269056.1 | 3 | 1310 | |||
| NM_001282128.1 | NP_001269057.1 | 3 | 1542 | |||
| NM_030178.2 | NP_084454.1 | 3 | 1310 | |||
| GenBank mRNA | AK043372.1 | - | 3 | 1543 | ||
| AK156375.1 | - | 3 | 1490 | |||
| BC031594.1 | - | 1 | 216 | |||
| BC046521.1 | - | 3 | 1250 | 5363697 | ||
| BC138361.1 | - | 3 | 1134 | 8861381 | ||
| BC145256.1 | - | 3 | 1366 | 9053809 | ||
| DQ288860.1 | - | 2 | 1099 | |||

Brpf1
Genomic Map