siRNA Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114361.1 | NP_001107833.1 | 8, 9 | 1114 | ||
| NM_001114362.1 | NP_001107834.1 | 6, 7 | 907 | |||
| NM_001286567.1 | NP_001273496.1 | 8, 9 | 1017 | |||
| NM_199466.3 | NP_955760.3 | 5, 6 | 778 | |||
| GenBank mRNA | AK144604.1 | - | 8, 9 | 1115 | ||
| BC065178.1 | - | 8, 9 | 955 | 6827397 | ||
| BC066099.1 | - | 5, 6 | 709 | 30533573 | ||
| BC067011.1 | - | 5, 6 | 727 | 6405184 | ||
| BC070427.1 | - | 6, 7 | 847 | 6413041 | ||
| BC079619.1 | - | 5, 6 | 709 | 30533573 | ||
| DQ230326.1 | - | 7, 8 | 797 | |||

Eml4
Genomic Map