siRNA Details
Gene Symbol: NKX2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002509.3 | NP_002500.1 | 2 | 1730 | ||
XM_006723566.3 | XP_006723629.1 | 2 | 1387 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NKX2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002509.3 | NP_002500.1 | 2 | 1730 | ||
XM_006723566.3 | XP_006723629.1 | 2 | 1387 |